siRNA Details
| Gene Symbol: PSMD6-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038286.1 | 1 | 81 | |||
| GenBank mRNA | AK023371.1 | - | 1 | 81 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000472046.1 | - | 1 | 81 | Chr. 3 - 64004022 - 64012148 [+] on Build GRCh38 | |

PSMD6-AS2
Genomic Map