siRNA Details
| Gene Symbol: LOC105374546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134673.1 | 2 | 471 | |||
| NR_134674.1 | 2 | 302 | ||||
| NR_134675.1 | 2 | 319 | ||||
| GenBank mRNA | BX100535.1 | - | 2 | 471 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374546| Gene Symbol: LOC105374546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134673.1 | 2 | 471 | |||
| NR_134674.1 | 2 | 302 | ||||
| NR_134675.1 | 2 | 319 | ||||
| GenBank mRNA | BX100535.1 | - | 2 | 471 | ||