siRNA Details
| Gene Symbol: LOC105374546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134673.1 | 2 | 332 | |||
| NR_134674.1 | 2 | 163 | ||||
| NR_134675.1 | 2 | 180 | ||||
| GenBank mRNA | BU942553.1 | - | 2 | 148 | ||
| BX100535.1 | - | 2 | 332 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374546| Gene Symbol: LOC105374546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134673.1 | 2 | 332 | |||
| NR_134674.1 | 2 | 163 | ||||
| NR_134675.1 | 2 | 180 | ||||
| GenBank mRNA | BU942553.1 | - | 2 | 148 | ||
| BX100535.1 | - | 2 | 332 | |||