siRNA Details
| Gene Symbol: TM4SF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046650.1 | 1 | 133 | |||
| NR_109809.1 | 1 | 133 | ||||
| NR_109810.1 | 1 | 133 | ||||
| GenBank mRNA | CB956551.1 | - | 1 | 136 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000496491.1 | - | 1 | 133 | Chr. 3 - 149377778 - 149386583 [+] on Build GRCh38 | |

TM4SF1-AS1
Genomic Map