siRNA Details
Gene Symbol: TM4SF1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046650.1 | 1 | 294 | |||
NR_109809.1 | 1 | 294 | ||||
NR_109810.1 | 1 | 294 | ||||
GenBank mRNA | CB956551.1 | - | 1 | 297 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000496491.1 | - | 1 | 294 | Chr. 3 - 149377778 - 149386583 [+] on Build GRCh38 |
