siRNA Details
| Gene Symbol: LINC01168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046231.1 | 3 | 4592 | |||
| GenBank mRNA | AK125849.1 | - | 3 | 4592 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000461291.2 | - | 3 | 4592 | Chr. 10 - 132965534 - 132976354 [+] on Build GRCh38 | |

LINC01168
Genomic Map