siRNA Details
Gene Symbol: LINC01168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046231.1 | 2 | 2220 | |||
GenBank mRNA | AK125849.1 | - | 2 | 2220 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000461291.2 | - | 2 | 2220 | Chr. 10 - 132965534 - 132976354 [+] on Build GRCh38 |
