siRNA Details
| Gene Symbol: LOC100507642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108064.1 | 3 | 659 | |||
| NR_108065.1 | 2 | 552 | ||||
| GenBank mRNA | AK024243.1 | - | 2 | 3088 | ||
| AK310146.1 | - | 2 | 526 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507642| Gene Symbol: LOC100507642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108064.1 | 3 | 659 | |||
| NR_108065.1 | 2 | 552 | ||||
| GenBank mRNA | AK024243.1 | - | 2 | 3088 | ||
| AK310146.1 | - | 2 | 526 | |||