siRNA Details
| Gene Symbol: LINC00886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038387.1 | 3 | 2377 | |||
| GenBank mRNA | AK056401.1 | - | 3 | 2329 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000472943.5 | - | 3 | 2349 | Chr. 3 - 156747346 - 156817034 [-] on Build GRCh38 | |

LINC00886
Genomic Map