siRNA Details
Gene Symbol: LSAMP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109998.1 | 2 | 115 | |||
GenBank mRNA | BX110108.1 | - | 2 | 115 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000490351.1 | - | 2 | 115 | Chr. 3 - 116360024 - 116370090 [+] on Build GRCh38 |
