siRNA Details
| Gene Symbol: LSAMP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109998.1 | 4 | 333 | |||
| GenBank mRNA | BX110108.1 | - | 4 | 333 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000490351.1 | - | 4 | 333 | Chr. 3 - 116360024 - 116370090 [+] on Build GRCh38 | |

LSAMP-AS1
Genomic Map