siRNA Details
| Gene Symbol: LOC100506489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC039665.1 | - | 2 | 878 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000470532.1 | - | 2 | 878 | Chr. 7 - 115679345 - 115682618 [-] on Build GRCh38 | |

LOC100506489
Genomic Map