siRNA Details
| Gene Symbol: PRICKLE2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045697.1 | 3 | 3872 | |||
| GenBank mRNA | AL832189.1 | - | 3 | 3872 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000482609.1 | - | 3 | 3872 | Chr. 3 - 64067964 - 64103131 [+] on Build GRCh38 | |

PRICKLE2-AS1
Genomic Map