siRNA Details
| Gene Symbol: LOC100507461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134937.1 | 2 | 322 | |||
| GenBank mRNA | AA393442.1 | - | 2 | 324 | ||
| BX096822.1 | - | 2 | 277 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000490465.1 | - | 2 | 322 | Chr. 3 - 148078159 - 148088029 [+] on Build GRCh38 | |

LOC100507461
Genomic Map