siRNA Details
| Gene Symbol: LINC00877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104116.1 | 3 | 654 | |||
| GenBank mRNA | AK097190.1 | - | 3 | 626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00877| Gene Symbol: LINC00877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104116.1 | 3 | 654 | |||
| GenBank mRNA | AK097190.1 | - | 3 | 626 | ||