siRNA Details
Gene Symbol: LOC101928211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110180.1 | 4 | 494 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000470677.1 | - | 3 | 184 | Chr. 7 - 124337380 - 124349860 [-] on Build GRCh38 |
