siRNA Details
Gene Symbol: LOC101927494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135563.1 | 2 | 947 | |||
GenBank mRNA | BC035252.1 | - | 2 | 947 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000460586.1 | - | 2 | 947 | Chr. 3 - 86481943 - 86496996 [+] on Build GRCh38 |
