siRNA Details
| Gene Symbol: LINC01487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125399.1 | 2 | 158 | |||
| GenBank mRNA | BC037382.1 | - | 1 | 657 | ||
| BU570602.1 | - | 2 | 158 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000462531.1 | - | 1 | 657 | Chr. 3 - 155240945 - 155243124 [+] on Build GRCh38 | |

LINC01487
Genomic Map