siRNA Details
| Gene Symbol: LOC105375483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133947.1 | 1 | 21 | |||
| GenBank mRNA | BC041947.1 | - | 1 | 21 | ||
| DA346765.1 | - | 1 | 21 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000472838.1 | - | 1 | 21 | Chr. 7 - 123994622 - 124027659 [+] on Build GRCh38 | |

LOC105375483
Genomic Map