siRNA Details
Gene Symbol: LINC01192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033945.1 | 2 | 134 | |||
GenBank mRNA | AA382967.1 | - | 2 | 117 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000470546.5 | - | 2 | 134 | Chr. 3 - 163177243 - 163303301 [-] on Build GRCh38 |
