siRNA Details
| Gene Symbol: LINC00635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015414.1 | 6 | 561 | |||
| GenBank mRNA | BC045586.1 | - | 6 | 561 | ||
| BC101990.1 | - | 2 | 155 | |||
| BC101991.1 | - | 2 | 155 | |||
| BC104745.1 | - | 2 | 155 | |||
| BC104746.1 | - | 2 | 155 | |||
| BC113056.1 | - | 2 | 155 | |||
| BC113057.1 | - | 2 | 155 | |||

LINC00635
Genomic Map