siRNA Details
| Gene Symbol: LOC105374177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135546.1 | 1 | 737 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000469196.1 | - | 1 | 737 | Chr. 3 - 157081841 - 157088547 [+] on Build GRCh38 | |

LOC105374177
Genomic Map