siRNA Details
| Gene Symbol: LOC100505609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132416.1 | 2 | 408 | |||
| GenBank mRNA | BC041442.1 | - | 2 | 407 | ||
| BI830672.1 | - | 2 | 409 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000488262.1 | - | 2 | 407 | Chr. 3 - 180414174 - 180422506 [+] on Build GRCh38 | |

LOC100505609
Genomic Map