siRNA Details
| Gene Symbol: LINC00893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027455.5 | 10 | 1714 | |||
| GenBank mRNA | AK124040.1 | - | 10 | 1714 | ||
| U66044.1 | - | 4 | 475 | |||
| U66045.1 | - | 4 | 475 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00893| Gene Symbol: LINC00893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027455.5 | 10 | 1714 | |||
| GenBank mRNA | AK124040.1 | - | 10 | 1714 | ||
| U66044.1 | - | 4 | 475 | |||
| U66045.1 | - | 4 | 475 | |||