siRNA Details
| Gene Symbol: CECR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038398.2 | 3 | 668 | |||
| GenBank mRNA | AF277398.1 | - | 3 | 668 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000428828.1 | - | 3 | 668 | Chr. 22 - 17256859 - 17266733 [-] on Build GRCh38 | |

CECR3
Genomic Map