siRNA Details
Gene Symbol: LOC101929159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135550.1 | 2 | 487 | |||
GenBank mRNA | BC044922.2 | - | 2 | 487 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000465933.1 | - | 2 | 487 | Chr. 3 - 57693205 - 57696596 [+] on Build GRCh38 |
