siRNA Details
| Gene Symbol: LOC101929159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135550.1 | 2 | 510 | |||
| GenBank mRNA | BC044922.2 | - | 2 | 510 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000465933.1 | - | 2 | 510 | Chr. 3 - 57693205 - 57696596 [+] on Build GRCh38 | |

LOC101929159
Genomic Map