siRNA Details
Gene Symbol: LOC102546299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105065.1 | 1 | 65 | |||
GenBank mRNA | BX112058.1 | - | 1 | 65 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000522762.1 | - | 1 | 65 | Chr. 5 - 164470279 - 164542984 [+] on Build GRCh38 |
