siRNA Details
| Gene Symbol: LOC105376382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134491.1 | 2 | 336 | |||
| GenBank mRNA | BF218358.1 | - | 2 | 337 | ||
| BG035368.1 | - | 313 | ||||
| CF130727.1 | - | 2 | 310 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000478294.1 | - | 2 | 336 | Chr. 10 - 5594991 - 5596118 [-] on Build GRCh38 | |

LOC105376382
Genomic Map