siRNA Details
| Gene Symbol: LINC00996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034033.1 | 4 | 1617 | |||
| GenBank mRNA | AK097158.1 | - | 4 | 1519 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000477392.1 | - | 4 | 1617 | Chr. 7 - 150433654 - 150448140 [+] on Build GRCh38 | |

LINC00996
Genomic Map