siRNA Details
| Gene Symbol: LOC100507661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109989.1 | 5 | 731 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000484765.2 | - | 4 | 696 | Chr. 3 - 168903366 - 168921996 [+] on Build GRCh38 | |

LOC100507661
Genomic Map