siRNA Details
Gene Symbol: LOC100507661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109989.1 | 4 | 375 | |||
GenBank mRNA | BX118401.1 | - | 4 | 375 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100507661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109989.1 | 4 | 375 | |||
GenBank mRNA | BX118401.1 | - | 4 | 375 |