siRNA Details
| Gene Symbol: KCNAB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046618.2 | 1, 2 | 150 | |||
| GenBank mRNA | BX104028.1 | - | 1, 2 | 150 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000467794.1 | - | 1, 2 | 119 | Chr. 3 - 156441157 - 156446905 [-] on Build GRCh38 | |

KCNAB1-AS1
Genomic Map