siRNA Details
| Gene Symbol: LINC00901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121607.1 | 2 | 1169 | |||
| GenBank mRNA | BC040587.1 | - | 2 | 1169 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000487512.1 | - | 2 | 1169 | Chr. 3 - 116921431 - 116932238 [+] on Build GRCh38 | |

LINC00901
Genomic Map