siRNA Details
Gene Symbol: LINC00901 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121607.1 | 2 | 878 | |||
GenBank mRNA | BC040587.1 | - | 2 | 878 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000487512.1 | - | 2 | 878 | Chr. 3 - 116921431 - 116932238 [+] on Build GRCh38 |
