siRNA Details
Gene Symbol: LOC101927866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135560.1 | 3 | 380 | |||
NR_135561.1 | 2 | 356 | ||||
GenBank mRNA | BX537734.1 | - | 2 | 316 | ||
DA114969.1 | - | 3 | 381 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000471638.1 | - | 2 | 316 | Chr. 3 - 146921963 - 146925218 [+] on Build GRCh38 |
