siRNA Details
Gene Symbol: LOC101927550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110102.1 | 1 | 776 | |||
GenBank mRNA | AK001533.1 | - | 1 | 775 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000360902.1 | - | 1 | 776 | Chr. 7 - 99013165 - 99036477 [+] on Build GRCh38 |
