siRNA Details
Gene Symbol: LOC105374928 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038755.1 | - | 2 | 157 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000423149.1 | - | 2 | 161 | Chr. 6 - 11487226 - 11515710 [+] on Build GRCh38 |
