siRNA Details
| Gene Symbol: TUSC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015391.1 | 2 | 777 | |||
| GenBank mRNA | BC035185.2 | - | 2 | 786 | ||
| BC104179.1 | - | 2 | 596 | |||
| BC104180.1 | - | 2 | 596 | |||
| BC112981.1 | - | 2 | 596 | |||
| BC112982.1 | - | 2 | 596 | |||

TUSC7
Genomic Map