siRNA Details
| Gene Symbol: LINC01510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120506.1 | 5 | 1111 | |||
| GenBank mRNA | BG189453.1 | - | 2 | 446 | ||
| BG195638.1 | - | 2 | 445 | |||
| BG213277.1 | - | 2 | 448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01510| Gene Symbol: LINC01510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120506.1 | 5 | 1111 | |||
| GenBank mRNA | BG189453.1 | - | 2 | 446 | ||
| BG195638.1 | - | 2 | 445 | |||
| BG213277.1 | - | 2 | 448 | |||