siRNA Details
| Gene Symbol: LINC01510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120506.1 | 5 | 1345 | |||
| GenBank mRNA | BG189453.1 | - | 2 | 680 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000421965.1 | - | 2 | 679 | Chr. 7 - 116563594 - 116571234 [-] on Build GRCh38 | |

LINC01510
Genomic Map