siRNA Details
| Gene Symbol: LOC105374060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135547.1 | 4 | 414 | |||
| GenBank mRNA | BX092438.1 | - | 4 | 294 | ||
| EU250752.1 | - | 4 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374060| Gene Symbol: LOC105374060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135547.1 | 4 | 414 | |||
| GenBank mRNA | BX092438.1 | - | 4 | 294 | ||
| EU250752.1 | - | 4 | 415 | |||