siRNA Details
| Gene Symbol: LOC101926983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135571.1 | 2 | 190 | |||
| GenBank mRNA | BC032918.2 | - | 2 | 193 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000490647.1 | - | 2 | 193 | Chr. 3 - 120833440 - 120836360 [-] on Build GRCh38 | |

LOC101926983
Genomic Map