siRNA Details
Gene Symbol: LINC00880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034007.1 | 4 | 1007 | |||
GenBank mRNA | BC040669.1 | - | 4 | 1002 | ||
BC122546.1 | - | 1 | 607 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000471357.1 | - | 4 | 1009 | Chr. 3 - 157081667 - 157123004 [-] on Build GRCh38 |
