siRNA Details
| Gene Symbol: LOC100507389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038455.1 | 5 | 1475 | |||
| GenBank mRNA | AK096143.1 | - | 5 | 1476 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000476385.1 | - | 5 | 1475 | Chr. 3 - 142926675 - 142942534 [+] on Build GRCh38 | |

LOC100507389
Genomic Map