siRNA Details
| Gene Symbol: LOC100507210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135574.1 | 2 | 346 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000483245.1 | - | 2 | 342 | Chr. 3 - 134055256 - 134057648 [-] on Build GRCh38 | |

LOC100507210
Genomic Map