siRNA Details
| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055475.1 | - | 3 | 1070 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000430883.5 | - | 3 | 1070 | Chr. 22 - 37339583 - 37385283 [-] on Build GRCh38 | |

ELFN2
Genomic Map