siRNA Details
Gene Symbol: LOC105374313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136187.1 | 2 | 229 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000487840.5 | - | 2 | 120 | Chr. 3 - 150039214 - 150213614 [+] on Build GRCh38 |
