siRNA Details
| Gene Symbol: LOC105374313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136187.1 | 2 | 176 | |||
| GenBank mRNA | BX091828.1 | - | 2 | 176 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000487840.5 | - | 2 | 67 | Chr. 3 - 150039214 - 150213614 [+] on Build GRCh38 | |

LOC105374313
Genomic Map