siRNA Details
Gene Symbol: LOC101929371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109861.1 | 2 | 666 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000449270.1 | - | 2 | 911 | Chr. 20 - 9986088 - 10007116 [+] on Build GRCh38 |
